Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin F Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Cyclin F |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Cyclin F Polyclonal specifically detects Cyclin F in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cyclin F | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
899 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GDQESEGEKEGDVTAPSGILDVTVVYLNPEQHCCQESSDEEACPEDKGPQDPQALALDTQIPATPGPKPLVR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Unconjugated | |
RUO | |
Human | |
cyclin F, cyclin-F, FBX1FBXO1F-box only protein 1, G2/mitotic-specific cyclin-F | |
CCNF | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title