Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566341 Supplier Invitrogen™ Supplier No. PA566341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRQVSLTAVKQ RFEDKRYGEI SQEEVLSYSQ LCAALGVTQD SPDPPTFLST GEIHAFLSSP SGRRTKRKRE N Highest antigen sequence identity to the following orthologs - mouse 78%, rat 82%.

This gene encodes a member of the cyclin family. Cyclins are important regulators of cell cycle transitions through their ability to bind and activate cyclin-dependent protein kinases. This member also belongs to the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it was one of the first proteins in which the F-box motif was identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin F
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNF
Gene Accession No. P41002
Gene Alias CCNF; CDK2; CycF; cyclin F; cyclin-F; EC 2.7.11.22; F-box only protein 1; FBX1; Fbxo1; G2/mitotic-specific cyclin-F; P33(CDK2)
Gene Symbols CCNF
Host Species Rabbit
Immunogen Recombinant Human Cyclin F. Recombinant protein control fragment (Product #RP-107023).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.