Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclin F Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567186
Description
Immunogen sequence: GDQESEGEKEG DVTAPSGILD VTVVYLNPEQ HCCQESSDEE ACPEDKGPQD PQALALDTQI PATPGPKPLV R Highest antigen sequence identity to the following orthologs - mouse 69%, rat 69%.
This gene encodes a member of the cyclin family. Cyclins are important regulators of cell cycle transitions through their ability to bind and activate cyclin-dependent protein kinases. This member also belongs to the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it was one of the first proteins in which the F-box motif was identified.
Specifications
Cyclin F | |
Polyclonal | |
Unconjugated | |
CCNF | |
CCNF; CDK2; CycF; cyclin F; cyclin-F; EC 2.7.11.22; F-box only protein 1; FBX1; Fbxo1; G2/mitotic-specific cyclin-F; P33(CDK2) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
899 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P41002 | |
CCNF | |
Recombinant Human Cyclin F. Recombinant protein control fragment (Product #RP-107826). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction