Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin G1 Polyclonal Antibody, Abbomax

Catalog No. 502203532
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3532 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3532 Supplier Abbomax Inc Supplier No. 50010684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF7 Cellular location: Nuclear.

Cyclin G1 Polyclonal antibody specifically detects Cyclin G1 in Human samples. It is validated for Western Blot

Specifications

Antigen Cyclin G1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Ccng1
Gene Accession No. P51959
Gene Alias AI314029; CCNG; CCNG1; CYCG; CYCG1; cyclin G1; cyclin-G; Cyclin-G1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminal of human cyclin G1
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.