Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin H Antibody (3G6), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259440DL594
This item is not returnable.
View return policy
Description
Cyclin H Monoclonal antibody specifically detects Cyclin H in Human samples. It is validated for Western Blot, ELISASpecifications
Cyclin H | |
Monoclonal | |
DyLight 594 | |
CAK, cyclin H, cyclin-dependent kinase-activating kinase complex subunit, cyclin-H, MO15-associated protein, p34CAK complex subunit, p37CDK-activating kinase complex subunit | |
Recombinant human Cyclin H (1-323aa) purified from E.coli | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
902 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
3G6 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction