Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin H Mouse anti-Human, Clone: 3G6, Novus Biologicals™

Mouse Monoclonal Antibody
$342.00 - $499.50
Specifications
Antigen | Cyclin H |
---|---|
Clone | 3G6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1046, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25944050
![]() |
Novus Biologicals
NBP25944050UL |
50 μL |
Each for $342.00
|
|
|||||
NBP259440X
![]() |
Novus Biologicals
NBP259440100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
Cyclin H Monoclonal specifically detects Cyclin H in Human samples. It is validated for Western Blot, ELISA.Specifications
Cyclin H | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
902 | |
Recombinant human Cyclin H (1-323aa) purified from E.coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
3G6 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1046, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAK, cyclin H, cyclin-dependent kinase-activating kinase complex subunit, cyclin-H, MO15-associated protein, p34CAK complex subunit, p37CDK-activating kinase complex subunit | |
CCNH | |
IgG2b κ | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title