Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540668 Supplier Invitrogen™ Supplier No. PA540668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TTTTTTGGIL IGDRLYSEVS LTIDHSLIPE ERLSPTPSMQ DGLDLPSETD Sequence homology: Cow: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.

FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintenance of healthy cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin L1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ccnl1
Gene Accession No. Q9UK58
Gene Alias 2610030E23Rik; ANIA6A; ania-6a; AU018493; BM-001; Ccn1; Ccnl; CCNL1; Cyclin Ania-6a; cyclin L ania-6a; cyclin L gamma; cyclin L1; cyclin-L; Cyclin-L1; LOW QUALITY PROTEIN: cyclin-L1; PRO1073; UNQ530/PRO1073
Gene Symbols Ccnl1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCNL1 (aa 32-81).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57018
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.