Learn More
Invitrogen™ Cyclin L1 Polyclonal Antibody

Description
Cyclin L1 Polyclonal Antibody detects endogenous levels of Cyclin L1 protein.
Specifications
Specifications
| Antigen | Cyclin L1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ccnl1 |
| Gene Accession No. | Q52KE7, Q9R1Q2, Q9UK58 |
| Gene Alias | 2610030E23Rik; ANIA6A; ania-6a; AU018493; BM-001; Ccn1; Ccnl; CCNL1; Cyclin Ania-6a; cyclin L ania-6a; cyclin L gamma; cyclin L1; cyclin-L; Cyclin-L1; LOW QUALITY PROTEIN: cyclin-L1; PRO1073; UNQ530/PRO1073 |
| Gene Symbols | Ccnl1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.