Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin L2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10087009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10087009 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10087009 Supplier Novus Biologicals Supplier No. NB10087009
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Cyclin L2 Polyclonal specifically detects Cyclin L2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunoblotting, Knockdown Validated.

Specifications

Antigen Cyclin L2
Applications Western Blot, Immunoprecipitation, Immunoblot, KnockDown
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate, Immunoblotting, Knockdown Validated
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q96S94
Gene Alias ania-6b, CCNM, cyclin L2, cyclin M, cyclin-L2, DKFZp761A1210, DKFZp762O195, HCLA-ISO, HLA-ISO, Paneth cell-enhanced expression protein, PCEE, SB138
Gene Symbols CCNL2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 470 and 520 of human cyclin L2 using the numbering given in entry NP_112199.2 (GeneID 81669).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81669
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.