Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100708 Supplier Invitrogen™ Supplier No. PA5100708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCNL2.

CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin L2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCNL2
Gene Accession No. Q5I0H5, Q96S94, Q9JJA7
Gene Alias 1700010A01Rik; 1810019L15Rik; 2010319M22Rik; AA409936; Ania6b; ANIA-6B; AW261738; Ccnl2; CCNM; CCNS; Cyclin Ania-6b; cyclin L2; cyclin M; cyclin S; cyclin-L2; HCLA-150; HCLA-ISO; HLA-ISO; hypothetical protein LOC538796; MNCb-5160; paneth cell enhanced expression; paneth cell-enhanced expression protein; PCEE; SB138
Gene Symbols CCNL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCNL2(Accession Q96S94), corresponding to amino acid residues K485-R520.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298686, 56036, 81669
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.