Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562738 Supplier Invitrogen™ Supplier No. PA562738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRLYSGVLIT LENCLLPDDK LRFTPSMSSG LDTDTETDLR VVGCEL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin L2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNL2
Gene Accession No. Q96S94
Gene Alias 1700010A01Rik; 1810019L15Rik; 2010319M22Rik; AA409936; Ania6b; ANIA-6B; AW261738; Ccnl2; CCNM; CCNS; Cyclin Ania-6b; cyclin L2; cyclin M; cyclin S; cyclin-L2; HCLA-150; HCLA-ISO; HLA-ISO; hypothetical protein LOC538796; MNCb-5160; paneth cell enhanced expression; paneth cell-enhanced expression protein; PCEE; SB138
Gene Symbols CCNL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cyclin L2. Recombinant protein control fragment (Product #RP-100477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.