Learn More
Cyclin L2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301677A
Description
The recommended shelf life for this product is 1 year from date of receipt.
CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.Specifications
| Cyclin L2 | |
| Polyclonal | |
| Unconjugated | |
| CCNL2 | |
| ania-6b, CCNM, CCNS, cyclin M, cyclin S, HCLA-ISO, HLA-ISO, paneth cell-enhanced expression protein, PCEE, SB138 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81669 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96S94 | |
| CCNL2 | |
| Between 470 and 520 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.