Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin L2Rabbit anti-Human, Mouse, Polyclonal, Rockland™
SDP

Catalog No. 501991832
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1832 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1832 Supplier Rockland Immunochemicals Supplier No. 600401878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human Cyclin L2a and shows negligible reactivity to Cyclin L2b or Cyclin L1 isoforms. A BLAST analysis was used to suggest reactivity with this protein from mouse based on 100% homology for the immunogen sequence. Cross reactivity with Cyclin L2a protein from rat and chicken (called Cyclin S) is expected, as this sequence shows ∼ 80% homology with the protein from these sources. Cross reactivity with Cyclin L2a homologues from other sources has not been determined.

CCNL2 belongs to the cyclin family, Cyclin L subfamily. CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. CCNL2 also modulates the expression of critical apoptotic factor, leading to cell apoptosis.

Specifications

Antigen Cyclin L2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CCNL2
Gene Accession No. Q96S94, Q9JJA7
Gene Alias 1700010A01Rik; 1810019L15Rik; 2010319M22Rik; AA409936; Ania6b; ANIA-6B; AW261738; BOS_16006; Ccnl2; CCNM; CCNS; Cyclin Ania-6b; cyclin L2; cyclin M; cyclin S; Cyclin-L2; HCLA-150; HCLA-ISO; HLA-ISO; LOC100346778; MNCb-5160; paneth cell enhanced expression; Paneth cell-enhanced expression protein; PCEE; SB138
Gene Symbols CCNL2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56036, 81669
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.