Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin M2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987591
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7591 Supplier Bioss Supplier No. BS6912R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNNM2 is a divalent metal cation transporter. CNNM2 mediates transport of divalent metal cations in an order of Mg2+ > Co2+ > Mn2+ > Sr2+ > Ba2+ > Cu2+ > Fe2+.

Specifications

Antigen Cyclin M2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNNM2
Gene Accession No. Q9H8M5
Gene Alias Acdp2; ancient conserved domain protein 2; Ancient conserved domain-containing protein 2; AU015877; AW048635; BOS_23483; Clp2; CNNM2; cyclin and CBS domain divalent metal cation transport mediator 2; cyclin M2; cyclin-like protein 2; Cyclin-M2; HOMG6; HOMGSMR; mACDP2; Metal transporter CNNM2
Gene Symbols CNNM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Cyclin M2/CNNM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54805
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.