Learn More
Cyclin M3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304066AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Probable metal transporter.Specifications
Cyclin M3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8NE01 | |
CNNM3 | |
Between 657 and 707 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CNNM3 | |
ACDP3, ancient conserved domain protein 3, ancient conserved domain-containing protein 3, cyclin-M3, metal transporter CNNM3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26505 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.