Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin O Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145141 Supplier Invitrogen™ Supplier No. PA5145141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTSLLIACKQ VEVHPPRVKQ LLALCCGAFS RQQLCNLECI VLHKLHFTLG APTISFFLEH FTHARVEAGQ AEASEALEAQ ALA Antigen sequence identity: Mouse 87%; Rat 87%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin O
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNO
Gene Accession No. P22674
Gene Alias C86987; CCNO; Ccnu; CILD29; cyclin domain containing; cyclin O; cyclin U; cyclin-like uracil-DNA glycosylase; cyclin-O; E130318K11; RGD1565217; UDG2; Ung2; uracil DNA glycosylase 2; uracil-DNA glycosylase 2
Gene Symbols CCNO
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 180-262. Recombinant protein control fragment (Product #RP-110157).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.