Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclin Y Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144898
Description
Immunogen sequence: DKYKDLRRSA RKRSASADNL TLPRW Antigen sequence identity: Mouse 84%; Rat 84%.
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
Specifications
Cyclin Y | |
Polyclonal | |
Unconjugated | |
CCNY | |
1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; Ccny; CFP1; Cyclin box protein 1; cyclin fold protein 1; cyclin Y; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; cyc-Y; RGD1565969; si:dkey-18n12.2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
219771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8ND76 | |
CCNY | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 311-335. Recombinant protein control fragment (Product #RP-110073). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction