Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin Y Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523644

Catalog No. PIPA523644


Only null left
Add to Cart

Description

Description

Cyclin Y Polyclonal Antibody for Western Blot, IHC (P)

Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin Y
Polyclonal
Unconjugated
CCNY
1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; Ccny; CFP1; Cyclin box protein 1; cyclin fold protein 1; cyclin Y; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; cyc-Y; RGD1565969; si:dkey-18n12.2
Rabbit
Antigen affinity chromatography, Protein A
RUO
219771, 67974
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q8BGU5, Q8ND76
CCNY
KLH conjugated synthetic peptide between 140-169 amino acids from the central region of human CCNY.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.