Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclin Y Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543110
Description
Peptide sequence: DENLHPLSKS EVPPDYDKHN PEQKQIYRFV RTLFSAAQLT AECAIVTLVY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
Specifications
| Cyclin Y | |
| Polyclonal | |
| Unconjugated | |
| CCNY | |
| 1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; Ccny; CFP1; Cyclin box protein 1; cyclin fold protein 1; cyclin Y; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y; cyc-Y; RGD1565969; si:dkey-18n12.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 219771 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8ND76 | |
| CCNY | |
| Synthetic peptide directed towards the middle region of human CCNY. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction