Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin Y Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562439
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2439 Supplier Bethyl Laboratories, Inc Supplier No. A302376A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.

Specifications

Antigen Cyclin Y
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CCNY
Gene Accession No. Q8ND76
Gene Alias C10orf9, CBCP1, CCNX, CFP1, cyc-Y, cyclin box protein 1, cyclin fold protein 1, cyclin-X, cyclin-Y, cyclin-box carrying protein 1
Gene Symbols CCNY
Host Species Rabbit
Immunogen Between 291 and 441
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219771
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.