Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutanecarbonitrile, 97%

Catalog No. p-7080675
Click to view available options
Quantity:
5 g
25 g

CAS: 4426-11-3 | C5H7N | 81.12 g/mol

It is used for research or analysis use only, as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for research or analysis use only, as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4426-11-3
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.12
MDL Number MFCD00013735
InChI Key GFBLFDSCAKHHGX-UHFFFAOYSA-N

Specifications

Density 0.868
Boiling Point 144°C to 146°C
Flash Point 42°C (107°F)
Odor Recognizable
Refractive Index 1.4325
Quantity 5 g
UN Number UN1992
Beilstein 2037573
Synonym cyanocyclobutane, cyclobutyl cyanide, cyclobutancarbonitrile, cyclobutane carbonitrile, acmc-1afpy, cyclobutanecarbonitrile 5g, cyanocyclobutane; cyclobutyl cyanide
Solubility Information Slightly soluble in water.
SMILES C1CC(C1)C#N
IUPAC Name cyclobutanecarbonitrile
PubChem CID 78148
Formula Weight 81.12
Percent Purity 97%
Chemical Name or Material Cyclobutanecarbonitrile
Show More Show Less
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 224-606-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.