Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Cyclodextrin, MP Biomedicals™

Catalog No. ICN19005205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19005205 5 g
ICN19005201 1 g
ICN19005225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19005205 Supplier MP Biomedicals, Inc Supplier No. 0219005205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 5 g
Formula Weight 972.9
Physical Form Powder
Chemical Name or Material α-Cyclodextrin
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.