Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, 99+%

Catalog No. p-7023036
Click to view available options
Quantity:
1000 mL
5000 mL

CAS: 110-82-7 | C6H12 | 84.16 g/mol

Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

Solubility
Miscible with ether, alcohol, acetone and olive oil. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.779
Boiling Point 80°C to 81°C
Quantity 1000 mL
Flash Point −18°C (−1°F)
Odor Pungent
Refractive Index 1.426
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
Solubility Information Miscible with ether,alcohol,acetone and olive oil. Immiscible with water.
Formula Weight 84.16
Percent Purity ≥99%
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category H225-H304-H315-H320-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXANE
EINECSNumber 203-806-2
RTECSNumber GU6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.