Cyclohexane, >99%, ACS, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10 Evaporation residue 0.002%, Substances darkened by sulfuric acid P.T., H2O 0.02%

Manufacturer: Alfa Aesar 22864AP

Quantity 500mL

 View more versions of this product

Catalog No. AA22864AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-82-7
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula C6H12
    Formula Weight 84.16
    MDL Number MFCD00003814
    Chemical Name or Material Cyclohexane, For ACS analysis
    Grade ACS Grade

    Specifications

    Formula Weight 84.16
    Quantity 500mL
    Color Colorless
    DOT Information Hazard Class: 3, Packing Group: II
    Density 0.779g/cm3
    EINECS Number 203-806-2
    Physical Form Liquid
    Ignition Point 245°C (473°F)
    Odor Pungent
    Appearance Colorless liquid
    Assay Percent Range >99%
    Molecular Formula C6H12
    MDL Number MFCD00003814
    CAS 110-82-7
    Health Hazard 1 H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.
    Chemical Name or Material Cyclohexane, For ACS analysis
    Grade ACS Grade

    H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit