Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, 99+%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.

Catalog No. p-217368
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Absorbance 0.01 max. at 300nm, 0.01 max. at 400nm, 0.02 max. at 260nm, 0.03 max. at 250nm, 0.08 max. at 240nm, 0.2 max. at 230nm, 0.5 max. at 220nm, 1 max. at 210nm
Density 0.7700g/mL
Boiling Point 81.0°C
Quantity 500 mL
Flash Point −18°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4250 to 1.4270
Residue after Evaporation 0.002% max.
Assay Percent Range 99+%
Specific Gravity 0.77
Linear Formula C6H12
Viscosity 0.94 mPa.s (20°C)
Beilstein 05, 20
Water 0.02% max. (Coulometric)
Merck Index 15, 2716
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 203-806-2
RTECSNumber GU6300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.