Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclohexane, Reagent, ACS, 99%, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
Description
Spectrum™ Chemical Cyclohexane, Reagent, ACS is used as a nonpolar solvent, in the calibration of differential scanning calorimetry and other industrial applications including a chemical intermediate. The distinctive detergent-like odor of cyclohexane is reminiscent of cleaning product and is sometimes used in cleaning products as a fragrance enhancer.
Specifications
CAS Max % | 100% |
UN Number | UN1145 |
Percent Purity | 99% |
Grade | ACS |
Quantity | 4 x 4 L |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 0.002% |
Water | 0.02% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction