Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanecarboxaldehyde, 97%

Catalog No. aab2383714
Click to view available options
Quantity:
25 g

Catalog No. AAB2383714

Supplier: Thermo Scientific Chemicals B2383714

Only null left
Add to Cart

CAS: 2043-61-0 | C7H12O | 112.17 g/mol

Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with ethyl diazoacetate catalyzed by basic, nanoparticulate magnesium oxide leading to α-diazo-β-hydroxyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with eth
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-61-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001457
InChI Key KVFDZFBHBWTVID-UHFFFAOYSA-N
Synonym cyclohexanecarboxaldehyde, cyclohexanal, formylcyclohexane, cyclohexanealdehyde, cyclohexanaldehyde, cyclohexylcarboxaldehyde, 1-formylcyclohexane, hexahydrobenzaldehyde, cyclohexylformaldehyde, cyclohexane formaldehyde
PubChem CID 16275
IUPAC Name cyclohexanecarbaldehyde
SMILES O=CC1CCCCC1

Specifications

Melting Point 34°C to 35°C
Density 0.93
Boiling Point 161°C to 163°C
Flash Point 40°C (105°F)
Refractive Index 1.45
Quantity 25 g
UN Number UN1989
Beilstein 878306
Sensitivity Air and light sensitive
Solubility Information Soluble in chloroform,ether,alcohol. Slightly soluble in water.
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material Cyclohexanecarboxaldehyde
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 218-057-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.