Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanecarboxylic acid, 98%

Catalog No. AC146560010
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle

CAS: 98-89-5 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-89-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001461
InChI Key NZNMSOFKMUBTKW-UHFFFAOYSA-N
Synonym hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
PubChem CID 7413
ChEBI CHEBI:36096
IUPAC Name cyclohexanecarboxylic acid
SMILES C1CCC(CC1)C(=O)O

Specifications

Melting Point 30.0°C to 32.0°C
Color White
Density 1.0300g/mL
Boiling Point 232.0°C to 233.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620
Linear Formula C6H11CO2H
Quantity 1 kg
Beilstein 09, 7
Specific Gravity 1.03
Merck Index 15, 2717
Solubility Information Solubility in water: 0.201g/100mL (15 c). Other solubilities: soluble in alcohol and ether
Formula Weight 128.17
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Cyclohexanecarboxylic acid, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-711-3
RTECSNumber GU8370000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.