Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methoxide, pure, anhydrous powder

Catalog No. AC173125000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Catalog No. Quantity Packaging
AC173125000 500 g Glass bottle
AC173120050 5 g Glass bottle
AC173121000 100 g Glass bottle
AC173120025 2.5 kg Plastic Drum
4 options
This item is not returnable. View return policy

Catalog No. AC173125000

Supplier: Thermo Scientific Chemicals 173125000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 124-41-4 | CH3NaO | 54.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4 , 497-19-8 , 1310-73-2
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Alkalinity total alkalinity (as NaOCH3):> 99%
Melting Point 126.0°C
Color White-Yellow
Density 0.8
Packaging Glass bottle
Linear Formula CH3ONa
Quantity 500 g
UN Number 1431
Beilstein 01, 280
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15, 8775
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene
Formula Weight 54.02
Percent Purity ≥97% (NaOCH3)
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium methoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 204-699-5
RTECSNumber PC3570000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.