Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanol, 99%

Catalog No. p-7023732
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 108-93-0 | C6H12O | 100.161 g/mol

Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

Solubility
Miscible with ethanol, ethyl ether acetone, chloroform, ethyl acetate, linseed oil and benzene. Slightly miscible with cold water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, hydrogen peroxide and nitric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-93-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00003855
InChI Key HPXRVTGHNJAIIH-UHFFFAOYSA-N
Synonym cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol
PubChem CID 7966
ChEBI CHEBI:18099
IUPAC Name cyclohexanol
SMILES C1CCC(CC1)O

Specifications

Melting Point 23°C to 24°C
Density 0.951
Boiling Point 160°C to 161°C
Flash Point 67°C (152°F)
Assay Percent Range 99%
Refractive Index 1.465
Quantity 100 mL
Beilstein 906744
Merck Index 14,2725
Solubility Information Miscible with ethanol,ethyl ether acetone,chloroform,ethyl acetate,linseed oil and benzene. Slightly miscible with cold water.
Formula Weight 100.16
Percent Purity 99%
Chemical Name or Material Cyclohexanol
Show More Show Less
Hazard Category H227-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H332-H315-H335-H227
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-630-6
RTECSNumber GV7875000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.