Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone 2,4-dinitrophenylhydrazone, 99%

Catalog No. AAA1220303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
AAA1220303 1 g
AAA1220306 5 g
AAA1220314 25 g
3 options

Catalog No. AAA1220303

Supplier: Thermo Scientific Chemicals A1220303

Only null left
Add to Cart

CAS: 1589-62-4 | C12H14N4O4 | 278.268 g/mol

Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.

Solubility
Soluble in water (partly).

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1589-62-4
Molecular Formula C12H14N4O4
Molecular Weight (g/mol) 278.268
MDL Number MFCD00001658
InChI Key QLWXZRVOHCYKKK-UHFFFAOYSA-N
Synonym cyclohexanone 2,4-dinitrophenylhydrazone, 1-cyclohexylidene-2-2,4-dinitrophenyl hydrazine, cyclohexanone-2,4-dinitrophenylhydrazone, cyclohexanone, 2,4-dinitrophenyl hydrazone, cyclohexanone 2,4-dinitrophenyl hydrazone, n-cyclohexylideneamino-2,4-dinitroaniline, acmc-20alct, maybridge1_001626
PubChem CID 74117
IUPAC Name N-(cyclohexylideneamino)-2,4-dinitroaniline
SMILES C1CCC(=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-])CC1

Specifications

Melting Point 158°C to 160°C
Quantity 1 g
Solubility Information Soluble in water (partly).
Formula Weight 278.27
Percent Purity 99%
Chemical Name or Material Cyclohexanone 2,4-dinitrophenylhydrazone
EINECSNumber 216-458-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.