Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cyclohexene
SCI_ED

Catalog No. p-4416569 Shop All Fisherbrand Products
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Solution

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 110-83-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Color Colorless
Density 0.810g/cmsuperscript_3;
Quantity 500 mL
Packaging Solution
Assay Percent Range 0.9999
Linear Formula C6H12
Formula Weight 82.13
Chemical Name or Material Cyclohexene
ShelfLife 2 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.