Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexene (Contains 0.01% tert-Butylcresol as An Inhibitor/Reagent), Fisher Chemical

Catalog No. O21111 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O2111-1 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. O2111-1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 110-83-8 , 128-37-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00001539
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Boiling Range 81° to 85°C
Melting Point -104°C
Density 0.811g/cm³
Boiling Point 83°C
Flash Point −20°C
Assay Percent Range ≥98 %
Packaging Amber Glass
Linear Formula CHCH(CH2)3CH2
Quantity 1 L
UN Number 2256
Vapor Density 2.8g/mol
Vapor Pressure 75mmHg at 20°C
Formula Weight 82.15
Percent Purity ≥98%
Grade Reagent
Concentration or Composition (by Analyte or Components) Contains 0.01% tert-Butylcresol as an inhibitor
Physical Form Liquid
Chemical Name or Material Cyclohexene, Contains 0.01% tert-butylcresol as an inhibitor
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.