Learn More
Cyclohexene oxide, 98+%
Description
It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.
Solubility
Insoluble in water
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -40°C |
| Density | 0.972 |
| Boiling Point | 129°C to 130°C |
| Flash Point | 24°C (75°F) |
| Refractive Index | 1.452 |
| Quantity | 2500 g |
| UN Number | UN2920 |
| Beilstein | 383568 |
| Solubility Information | Insoluble in water |
| Formula Weight | 98.15 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H332-H311-H314-H335 |
| Hazard Statement | GHS H Statement H311-H314-H318-H226-H302-H332 Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
| EINECSNumber | 206-007-7 |
| RTECSNumber | RN7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.