Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cycloheximide, 90%

Click to view available options
Quantity:
1g
5g
66-81-9
C15H23NO4
281.352
MFCD00082346
YPHMISFOHDHNIV-FSZOTQKASA-N
3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
6197
CHEBI:27641
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
CC1CC(C(=O)C(C1)C(CC2CC(=O)NC(=O)C2)O)C
66-81-9
C15H23NO4
281.352
MFCD00082346
YPHMISFOHDHNIV-FSZOTQKASA-N
3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
6197
CHEBI:27641
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
CC1CC(C(=O)C(C1)C(CC2CC(=O)NC(=O)C2)O)C
  • A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis
UN Number UN2811
Beilstein 88868
Merck Index 14,2728
Solubility Information Soluble in water at 20mg/mL
Formula Weight 281.35
Physical Form Powder
Percent Purity 90%
Boiling Point 492°C
Color White to Cream
Melting Point 111°C to 116°C
Quantity 1g
Chemical Name or Material Cycloheximide
Show More Show Less

GHS H Statement
H300-H360-H341
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : MA4375000