Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycloheximide from Microbial, MP Biomedicals

Catalog No. mp210018325
Click to view available options
Quantity:
25 g

  • Cycloheximide is used in protein synthesis in apoptosis
  • It is also used in plant research to study disease resistance and as an ethylene stimulant, useful in studies involving fruit and leaf production

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Melting Point 103°C to 115°C
Color White
Linear Formula C15H23NO4
Quantity 25 g
Formula Weight 281.4
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material CYCLOHEXIMIDE from Microbial
Recommended Storage +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.