Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Propionate 98.0+%, TCI America™
SDP

Catalog No. P050425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050425ML 25 mL
P0504500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P050425ML Supplier TCI America Supplier No. P050425ML

Chemical Identifiers

CAS 6222-35-1
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00046352
InChI Key MAMMVUWCKMOLSG-UHFFFAOYSA-N
Synonym Propionic Acid Cyclohexyl Ester
PubChem CID 61375
IUPAC Name cyclohexyl propanoate
SMILES CCC(=O)OC1CCCCC1

Specifications

Color Colorless
Boiling Point 193°C
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Propionate
EINECSNumber (3)-2359
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.