Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylamine, 0.50 Normal (N/2) in Methanol, Reagents

Catalog No. 01335554
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-554 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-554 Supplier Reagents Holdings Llc Supplier No. CC2678001C
Only null left
Add to Cart
Add to Cart

Cyclohexylamine, 0.50 Normal (N/2) in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-56-1 , 108-91-8
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Aminocyclohexane, Hexahydroaniline
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Boiling Point 65°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.7911
Grade Reagent
Physical Form Liquid
Chemical Name or Material Cyclohexylamine, 0.50 Normal (N/2) in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.