Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylamine, 98+%

Catalog No. AAA15851AP
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 108-91-8 | C6H13N | 99.177 g/mol

Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.

Solubility
Miscible with water, ethanol, ethers, acetone, alcohol and ketones.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, carbon dioxide, sodium hypochlorite, organic acids, mineral acids and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-91-8
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00001486
InChI Key PAFZNILMFXTMIY-UHFFFAOYSA-N
Synonym cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
PubChem CID 7965
ChEBI CHEBI:15773
IUPAC Name cyclohexanamine
SMILES C1CCC(CC1)N

Specifications

Melting Point -18°C
Density 0.868
Boiling Point 133°C to 134°C
Flash Point 27°C (81°F)
Refractive Index 1.4585
Quantity 500 mL
UN Number UN2357
Beilstein 471175
Sensitivity Air Sensitive
Merck Index 14,2729
Solubility Information Miscible with water,ethanol,ethers,acetone,alcohol and ketones.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material Cyclohexylamine
Show More Show Less
Hazard Category H226-H301+H311-H314-H361f
Hazard Statement GHS H Statement
H314-H226-H361-H302-H312
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CYCLOHEXYLAMINE
EINECSNumber 203-629-0
RTECSNumber GX0700000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.