Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclohexylurea, 98%

Click to view available options
Quantity:
25g
100g
500g
698-90-8
C7H14N2O
142.202
MFCD00014286
WUESWDIHTKHGQA-UHFFFAOYSA-N
1-cyclohexylurea,urea, cyclohexyl,n-cyclohexylurea,n-cyclohexlurea,cyclohexyl urea,amino-n-cyclohexylamide,cyclohexyl-urea,monocyclohexylurea,n'-cyclohexylurea,n'-cyclohexyl urea
69684
cyclohexylurea
C1CCC(CC1)NC(=O)N
698-90-8
C7H14N2O
142.202
MFCD00014286
WUESWDIHTKHGQA-UHFFFAOYSA-N
1-cyclohexylurea,urea, cyclohexyl,n-cyclohexylurea,n-cyclohexlurea,cyclohexyl urea,amino-n-cyclohexylamide,cyclohexyl-urea,monocyclohexylurea,n'-cyclohexylurea,n'-cyclohexyl urea
69684
cyclohexylurea
C1CCC(CC1)NC(=O)N
Melting Point 191°C to 195°C
Quantity 25g
Beilstein 636914
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Cyclohexylurea

RTECSNumber : YS7564300