Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylurea, 98%

Catalog No. AAB2491522
Click to view available options
Quantity:
25 g
100 g

CAS: 698-90-8 | C7H14N2O | 142.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-90-8
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD00014286
InChI Key WUESWDIHTKHGQA-UHFFFAOYSA-N
Synonym 1-cyclohexylurea, urea, cyclohexyl, n-cyclohexylurea, n-cyclohexlurea, cyclohexyl urea, amino-n-cyclohexylamide, cyclohexyl-urea, monocyclohexylurea, n'-cyclohexylurea, n'-cyclohexyl urea
PubChem CID 69684
IUPAC Name cyclohexylurea
SMILES C1CCC(CC1)NC(=O)N

Specifications

Melting Point 191°C to 195°C
Quantity 100 g
Beilstein 636914
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Cyclohexylurea
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 211-822-6
RTECSNumber YS7564300
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.