Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclon/CCDC86 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 149471AP150UL

Description
Coiled-coil domain-containing protein 86 (CCDC86) was identified as an immediate-early cytokine-responsive gene induced by interleukin 3 (IL-3) in a hematopoietic cell line. CCDC86 contains a coiled-coil domain in the C-terminus.Specifications
Cyclon/CCDC86 | |
Polyclonal | |
Unconjugated | |
CCDC86 | |
CCDC86, CYCLON | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108673, 293738, 79080 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XIB5, Q9H6F5, Q9JJ89 | |
CCDC86 | |
Cyclon/CCDC86 Fusion Protein Ag6754 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction