Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cycloolefin Microplate Lid
GREENER_CHOICE

Catalog No. p-7075502 Shop All Greiner Bio-One Products
Click to view available options
Sterility:
Non-sterile
Sterile

For use with 1536-Well Cycloolefin Low-Evaporation Clear Microplate.

  • Universal fit
  • Clear
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Cycloolefin
Sterility Non-sterile
Product Type Universal Lid
Plate Compatibility 1536-well Cycloolefin Low-evaporation Clear Microplate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.