Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cyclopamine
GREENER_CHOICESDP

Catalog No. 16231 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-231 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-231 Supplier Tocris Bioscience™ Supplier No. 1623/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of Hedgehog (Hh) signaling

Inhibitor of hedgehog (Hh) signaling, likely via direct inhibition of Smoothened, the accessory protein to the putative Hh receptor Patched. Anticancer and teratogenic in vivo. Depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

Chemical Identifiers

CAS 4449-51-8
Molecular Formula C27H41NO2

Specifications

Quantity 1 mg
Formula Weight 411.63
Purity >97%
Molecular Weight (g/mol) 411.63
Chemical Name or Material Cyclopamine
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.