Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Cyclopentadienylmanganese tricarbonyl
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 1g |
Sensitivity | Air sensitive |
Melting Point | 77°C |
UN Number | UN3466 |
Solubility Information | Soluble in water (70 ppm (25C) |
Formula Weight | 204.06 |
Physical Form | Crystalline |
Chemical Name or Material | Cyclopentadienylmanganese tricarbonyl |
Safety and Handling
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber : 235-142-4
TSCA : Yes
Recommended Storage : Ambient temperatures