Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclopentadienyltitanium trichloride, 99%, ACROS Organics™
Click to view available options
Quantity:
2.5g
10g
Packaging:
Glass bottle
Specifications
Color | Yellow to Orange |
Elemental Analysis | 98.5% min. |
Melting Point | 170°C |
Quantity | 2.5g |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Assay Percent Range | 99% |
Solubility Information | Solubility in water: reacts |
Formula Weight | 219.36 |
Physical Form | Crystals or Crystalline Powder |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RTECSNumber : XR1927300