Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentane, CHROMASOLV™, for HPLC, ≥75% cyclopentane basis, Solstice

Catalog No. 60045745 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-745 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-745 Supplier Solstice Supplier No. 2706011L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC,≥75% cyclopentane basis

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Absorbance max. 0.01 at 400 nm, max. 0.01 at 240 nm, max. 0.10 at 220 nm, max. 0.50 at 210 nm, max. 1.0 at 198 nm
CAS Max % 100.0000%
Melting Point -94°C (literature)
Density 0.751 g/mL at 25 °C(lit.)
Boiling Point 50°C (literature)
Flash Point −20°C
Packaging Glass Bottle
Linear Formula C5H10
Quantity 1 L
UN Number UN1146
Beilstein 1900195
Formula Weight 70.13
Percent Purity >75%
Grade HPLC
Chemical Name or Material Cyclopentane, CHROMASOLV™, for HPLC, ≥75% cyclopentane basis
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclopentane
EINECSNumber 206-016-6
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.