accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 287-92-3

IUPAC Name: cyclopentane
Synonyms: cyclopentane pentamethylene zyklopentan ciclopentano hsdb 62
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: RGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILES: C1CCCC1
Molecular Weight (g/mol) 
  • (6)
  • (12)
  • (7)
Boiling Point 
  • (6)
  • (4)
  • (1)
  • (3)
  • (1)
  • (7)
Quantity 
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (17)
  • (3)
Grade 
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (13)
  • (6)
  • (12)
  • (7)
  • (6)
  • (4)
  • (1)
  • (3)
  • (1)
  • (7)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (17)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)