Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentane, Spectrum™ Chemical
SDP

Catalog No. 18620377 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-377 200 L
18-620-374 500 mL
18-620-375 1 L
18-620-376 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-377 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1480200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Pentamethylene

Chemical Identifiers

CAS 287-92-3

Specifications

Boiling Point 49.2°C/120.56°F
Packaging Drum
Quantity 200 L
Molecular Weight (g/mol) 70.13
Grade Ungraded
Physical Form Liquid
Chemical Name or Material Cyclopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.