Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-chloropyridazine-4-carboxylate, 97%

Catalog No. AC432980050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 1093860-48-0 | C6H5ClN2O2 | 172.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093860-48-0
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD16660968
InChI Key QYQHGLKSLGUINZ-UHFFFAOYSA-N
Synonym 6-chloro-pyridazine-4-carboxylic acid methyl ester, methyl6-chloropyridazine-4-carboxylate, 4-pyridazinecarboxylicacid, 6-chloro-, methyl ester, 6-chloro-4-pyridazinecarboxylic acid, methyl ester, 4-pyridazinecarboxylic acid, 6-chloro-, methyl ester
PubChem CID 45790080
IUPAC Name methyl 6-chloropyridazine-4-carboxylate
SMILES COC(=O)C1=CC(Cl)=NN=C1

Specifications

Melting Point 88.4°C to 88.5°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 172.57
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl 6-chloropyridazine-4-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.